Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Furohydrazide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Furoic hydrazide2-Furoic acid Hydrazide2-Furoic acid hydrazide2-Furoylhydrazine2-Furancarboxylic acid Hydrazide2-Furoic Hydrazide2-Furancarboxylic acid hydrazide

A13630-30 A13630-18 A13630-30 A13630-18 A13630-09 A13630-09
AAA13630-18EA 193.99 USD InStock
CAAAAA13630-30 CAAAA13630-09 CAAAA13630-18 AAA13630-18 AAAA13630-30 AAA13630-09
2-Furohydrazide 98%
2-Furohydrazide


Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00003235
CAS Number: 3326-71-4
EINECS: 222-046-2
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty