Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Phenyl-2-pentenal mixture E/Z ≥90%

Supplier: Thermo Scientific Chemicals
Warning

A13635-14 A13635-22 A13635-22 A13635-06 A13635-14 A13635-06
AAA13635-14EA 197.99 USD InStock
AAA13635-14 AAAA13635-22 CAAAA13635-06 CAAAAA13635-22 AAA13635-06 CAAAA13635-14
2-Phenyl-2-pentenal mixture E/Z ≥90%
2-Phenyl-2-pentenal


Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Boiling Pt: 124…126 °C/14 mm
Density: 1,005
Flash Pt: >100 °C(212 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00051835
CAS Number: 3491-63-2
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty