Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Phenylpropionitrile 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: alpha-Methylphenylacetonitrile

A13636-09 A13636-14 A13636-18 A13636-09 A13636-14 A13636-18
AAA13636-18EA 167.52 USD InStock
CAAAAA13636-14 CAAAA13636-18 AAA13636-18 CAAAA13636-09 AAA13636-09 AAAA13636-14
2-Phenylpropionitrile 96%
2-Phenylpropionitrile


Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 230…232 °C
Density: 0.985
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001871
CAS Number: 1823-91-2
EINECS: 217-354-9
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty