Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethylamine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Diethylammonium chloride

A13637-0I A13637-30 A13637-0B A13637-30 A13637-0B A13637-0I
AAA13637-0IEA 323.99 USD InStock
CAAAA13637-0I AAA13637-0I AAA13637-0B CAAAA13637-30 CAAAA13637-0B AAA13637-30
Diethylamine hydrochloride 99%
Diethylamine hydrochloride


Formula: C₄H₁₂ClN
MW: 109.6 g/mol
Boiling Pt: 320…330 °C
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012499
CAS Number: 660-68-4
EINECS: 211-541-9
Merck Index: 13,03138
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty