Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Aspartic acid 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: DL-Aspartic acid2-Aminobutanedioic acid

A13646-0E A13646-0E A13646-36 A13646-36 A13646-22 A13646-22
AAA13646-36EA 59.99 USD InStock
AAA13646-36 AAA13646-22 AAA13646-0E CAAAA13646-22 CAAAA13646-36 CAAAA13646-0E
(±)-Aspartic acid 98+%
(±)-Aspartic acid


Formula: C₄H₇NO₄
MW: 133.10 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063083
CAS Number: 617-45-8
EINECS: 210-513-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty