Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Octylthiophene 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-n-Octylthiophene

A13672-06 A13672-18 A13672-18 A13672-14 A13672-14 A13672-06
AAA13672-14EA 272.99 USD InStock
AAA13672-14 AAA13672-06 AAAA13672-18 CAAAA13672-14 CAAAAA13672-18 CAAAA13672-06
2-Octylthiophene 98%
2-Octylthiophene


Formula: C₁₂H₂₀S
MW: 196.36 g/mol
Boiling Pt: 127…129 °C/12 mm
Density: 0.921
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041015
CAS Number: 880-36-4
EINECS: 212-913-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty