Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Difluoroaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

A13673-36 A13673-14 A13673-22 A13673-14 A13673-36 A13673-22
AAA13673-36EA 544.99 USD InStock
AAA13673-36 AAA13673-14 CAAAA13673-22 CAAAA13673-14 CAAAA13673-36 AAA13673-22
2,4-Difluoroaniline 99%
2,4-Difluoroaniline


Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 168…170 °C
Melting Pt: –8 °C to –7 °C
Density: 1.281
Flash Pt: 62 °C(143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007648
CAS Number: 367-25-9
EINECS: 206-687-5
UN: 2941
ADR: "6.1",III
Merck Index: 13,03171
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty