Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-N,N-diethylacetamide 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Chloro-n,n-diethylacetamideN,N,-Diethylchloroacetamide

A13691-22 A13691-14 A13691-22 A13691-14
AAA13691-14EA 86.99 USD InStock
CAAAA13691-22 CAAAA13691-14 AAA13691-14 AAA13691-22
2-Chloro-N,N-diethylacetamide 97%
2-Chloro-N,N-diethylacetamide


Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
Boiling Pt: 123…124 °C/15 mm
Density: 1.087
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000928
CAS Number: 2315-36-8
EINECS: 219-019-2
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty