Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: p-Hydroxybenzoic Acid

A13700-0B A13700-0I A13700-0I A13700-30 A13700-30 A13700-0B
AAA13700-30EA 37.48 USD InStock
CAAAA13700-30 AAA13700-30 CAAAA13700-0B AAA13700-0I CAAAA13700-0I AAA13700-0B
4-Hydroxybenzoic acid 99%
4-Hydroxybenzoic acid


Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 213…218 °C
Density: 1.398 g/cm³ (20 °C)
Flash Pt: 199 °C(390 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002547
CAS Number: 99-96-7
EINECS: 202-804-9
Merck Index: 13,04837
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty