Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adipic acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,6-Hexanedioic acid

A13705-0G A13705-0I A13705-0B A13705-0I A13705-0B A13705-0G
AAA13705-0BEA 39.69 USD InStock
AAA13705-0B CAAAA13705-0G CAAAA13705-0I CAAAA13705-0B AAA13705-0I AAA13705-0G
Adipic acid 99%
Adipic acid


Formula: HOOC(CH₂)₄COOH
MW: 146.14 g/mol
Boiling Pt: ca 205 °C/3 mm
Melting Pt: 151…155 °C
Density: 1.360
Flash Pt: 196 °C(384 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
Merck Index: 13,00163
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/30/2300230.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty