Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crotonoyl chloride 90% remainder mainly cis-isomer, tech., Technical Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Butenoyl chlorideCrotonic acid chloride

A13710-36 A13710-14 A13710-36 A13710-14 A13710-22 A13710-22
AAA13710-22EA 94.69 USD InStock
CAAAA13710-36 CAAAA13710-14 AAA13710-22 CAAAA13710-22 AAA13710-36 AAA13710-14
Crotonoyl chloride 90% remainder mainly cis-isomer, tech., Technical Grade
Crotonoyl chloride


Formula: C₄H₅ClO
MW: 104.53 g/mol
Boiling Pt: 121…123 °C
Density: 1.091
Flash Pt: 35 °C(95 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000734
CAS Number: 625-35-4
EINECS: 210-889-9
UN: 2920
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty