Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dibromosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3,5-Dibromo-2-hydroxybenzaldehyde

A13729-30 A13729-18 A13729-09 A13729-09 A13729-18 A13729-30
AAAA13729-30EA 1191.57 USD InStock
CAAAA13729-18 AAAA13729-30 CAAAAA13729-30 CAAAA13729-09 AAA13729-18 AAA13729-09
3,5-Dibromosalicylaldehyde 98%
3,5-Dibromosalicylaldehyde


Formula: C₇H₄Br₂O₂
MW: 279.92 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00003318
CAS Number: 90-59-5
EINECS: 202-003-4
Merck Index: 13,03052
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty