Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl(methyl)amine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-Benzylmethylamine

A13746-AK A13746-AE A13746-AU A13746-AE A13746-AU A13746-AK
AAAA13746-AEEA 30.6 USD InStock
AAAA13746-AE CAAAAA13746-AE AAA13746-AU CAAAA13746-AK CAAAA13746-AU AAA13746-AK
Benzyl(methyl)amine 97%
Benzyl(methyl)amine


Formula: C₆H₅CH₂NHCH₃
MW: 121.18 g/mol
Boiling Pt: 184…188 °C
Melting Pt: –55 °C
Density: 0.940
Flash Pt: 77 °C(170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008289
CAS Number: 103-67-3
EINECS: 203-133-4
UN: 2922
ADR: "8",III
https://us-legacy.vwr.com/stibo/web/std.lang.all/09/57/2370957.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty