Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(4-Fluorobenzoyl)propionic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Haloperidol Metabolite IIIR 11302

A13755-14 A13755-06 A13755-06 A13755-14
AAA13755-14EA 107.99 USD InStock
AAA13755-14 CAAAA13755-06 AAA13755-06 CAAAA13755-14
3-(4-Fluorobenzoyl)propionic acid 97%
3-(4-Fluorobenzoyl)propionic acid


Formula: C₁₀H₉FO₃
MW: 196.18 g/mol
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00002793
CAS Number: 366-77-8
EINECS: 206-679-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty