Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminobenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: p-Aminobenzophenone

A13757-09 A13757-09 A13757-18 A13757-18
AAA13757-18EA 267.99 USD InStock
CAAAA13757-09 AAA13757-18 CAAAA13757-18 AAA13757-09
4-Aminobenzophenone 98%
4-Aminobenzophenone


Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 245…247 °C/22 mm
Melting Pt: 121…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00007895
CAS Number: 1137-41-3
EINECS: 214-506-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty