Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2,4-dinitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Danger

A13774-22 A13774-36 A13774-0E A13774-22 A13774-0E A13774-36
AAA13774-22EA 53.31 USD InStock
CAAAA13774-22 AAA13774-22 AAA13774-36 AAA13774-0E CAAAA13774-0E CAAAA13774-36
1-Chloro-2,4-dinitrobenzene 98%
1-Chloro-2,4-dinitrobenzene


Formula: C₆H₃ClN₂O₄
MW: 202.55 g/mol
Boiling Pt: 314…316 °C
Melting Pt: 48…52 °C
Density: 1.700
Flash Pt: 186 °C(366 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007075
CAS Number: 97-00-7
EINECS: 202-551-4
UN: 3441
ADR: "6.1",II
Merck Index: 13,02155
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty