Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Iodohexane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Hexyl iodide

Stabilization: Stabilised with copper

A13784-36 A13784-14 A13784-22 A13784-22 A13784-36
AAA13784-36EA 318.99 USD InStock
CAAAA13784-22 AAA13784-36 AAA13784-14 AAA13784-22 CAAAA13784-36
1-Iodohexane 98% stabilized
1-Iodohexane


Formula: C₆H₁₃I
MW: 212.07 g/mol
Boiling Pt: 179…181 °C
Melting Pt: –75 °C
Density: 1.437
Flash Pt: 61 °C(141 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001102
CAS Number: 638-45-9
EINECS: 211-339-0
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty