Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

m-Toluic acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Methylbenzoic acid 3-Toluic acid

A13785-0B A13785-0I A13785-0B A13785-0I A13785-30 A13785-30
AAA13785-0IEA 326.59 USD InStock
AAA13785-0I CAAAA13785-30 AAA13785-0B CAAAA13785-0I CAAAA13785-0B AAA13785-30
m-Toluic acid 99%
m-Toluic acid


Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 263 °C
Melting Pt: 108…110 °C
Density: 1.050
Flash Pt: 150 °C(302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002523
CAS Number: 99-04-7
EINECS: 202-723-9
Merck Index: 12,09673
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty