Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caproic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hexanoic acid

A13789-0F A13789-AE A13789-0F A13789-AP A13789-AE
AAA13789-0FEA 153.99 USD InStock
AAA13789-0F AAAA13789-AE CAAAAA13789-AE CAAAA13789-0F AAA13789-AP
Caproic acid 98+%
Caproic acid


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 202…203 °C
Melting Pt: –4 °C
Density: 0.929
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004421
CAS Number: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: "8",III
Merck Index: 13,01765
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty