Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Toluic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methylbenzoic acid 2-Toluic acid

A13856-0B A13856-30 A13856-30 A13856-0I A13856-0I A13856-0B
AAA13856-0IEA 341.28 USD InStock
CAAAA13856-30 AAA13856-0I CAAAA13856-0I AAA13856-30 AAA13856-0B CAAAA13856-0B
o-Toluic acid 98+%
o-Toluic acid


Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 258…259 °C
Melting Pt: 103…106 °C
Density: 1.060
Flash Pt: 148 °C(298 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002477
CAS Number: 118-90-1
EINECS: 204-284-9
Merck Index: 12,09673
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty