Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,3',5,5'-Tetramethylbenzidine (TMB) 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: TMB

A13868-06 A13868-03 A13868-14 A13868-06 A13868-14 A13868-03
AAA13868-03EA 53.91 USD InStock
AAA13868-03 AAA13868-14 CAAAA13868-06 AAA13868-06 CAAAA13868-14 CAAAA13868-03
3,3',5,5'-Tetramethylbenzidine (TMB) 98%
3,3',5,5'-Tetramethylbenzidine (TMB)
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes; particularly useful in ELISA techniques, blue result can be read spectrophotometrically at 370 or 650 nm3,3',5,5'-Tetramethylbenzidine is used as a clinical reagent for testing blood.

It is also a chromogenic substrate which is used in staining procedures in immunohistochemistry. Further, it is used as a visualizing reagent in enzyme-linked immunosorbent assays. In addition to this, it is used as a replacement for carcinogenic compounds such as benzidine and o-phenylenediamine.


Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 168…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00007748
CAS Number: 54827-17-7
EINECS: 259-364-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty