Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methylvaleric acid 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (±)-2-Methylpentanoic acid

A13873-0E A13873-0E A13873-22 A13873-22
AAA13873-22EA 47.49 USD InStock
AAA13873-22 CAAAA13873-0E CAAAA13873-22 AAA13873-0E
(±)-2-Methylvaleric acid 98+%
(±)-2-Methylvaleric acid


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 196…197 °C
Melting Pt: –85 °C
Density: 0.930
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002671
CAS Number: 97-61-0
EINECS: 202-594-9
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty