Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyruvic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Ketopropionic acid 2-Oxopropionic acid α-Ketopropionic acid

A13875-22 A13875-0E A13875-36 A13875-0E A13875-22 A13875-36
AAA13875-36EA 215.99 USD InStock
AAA13875-36 CAAAA13875-0E AAA13875-0E AAA13875-22 CAAAA13875-36 CAAAA13875-22
Pyruvic acid 98%
Pyruvic acid


Formula: C₃H₄O₃
MW: 88.06 g/mol
Boiling Pt: 164…166 °C
Melting Pt: 11…12 °C
Density: 1.267
Flash Pt: 83 °C(181 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002585
CAS Number: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: "8",III
Merck Index: 12,08205
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty