Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylanisole 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methoxytolueneo-Cresol methyll ether2-Methylanisole1-Methoxy-2-methylbenzene

A13897-30 A13897-0B A13897-30 A13897-0B A13897-18 A13897-18
AAA13897-0BEA 814.82 USD InStock
CAAAA13897-18 AAA13897-0B CAAAA13897-30 AAA13897-18 AAA13897-30 CAAAA13897-0B
2-Methylanisole 99%
2-Methylanisole


Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 170…172 °C
Density: 0.985
Flash Pt: 51 °C(123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008373
CAS Number: 578-58-5
EINECS: 209-426-3
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty