Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: DL-Serine Methyl Ester HydrochlorideH-DL-Ser-OMe·HClmethyl 2-amino-3-hydroxypropanoateMethyl DL-serinate hydrochloride3-hydroxy-1-methoxy-1-oxopropan-2-aminium chloride

A13904-18 A13904-18 A13904-09 A13904-30 A13904-30 A13904-09
AAA13904-09EA 29.39 USD InStock
AAA13904-09 AAA13904-18 CAAAA13904-30 CAAAA13904-18 CAAAA13904-09 AAA13904-30
Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥98%
Methyl-2-amino-3-hydroxypropanoate hydrochloride


Formula: C₄H₁₀ClNO₃
MW: 155.578 g/mol
Melting Pt: ca 134 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012593
CAS Number: 5619-04-5
EINECS: 227-047-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty