Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tiglic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (E)-2-Methylbut-2-enoic acidtrans-2,3-Dimethylacrylic acid

A13905-36 A13905-36 A13905-22 A13905-22 A13905-14 A13905-14
AAA13905-22EA 87.59 USD InStock
CAAAA13905-22 CAAAA13905-36 AAA13905-22 CAAAA13905-14 AAA13905-36 AAA13905-14
Tiglic acid 98+%
Tiglic acid


Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 198…199 °C
Melting Pt: 62…66 °C
Density: 0.969
Flash Pt: 101 °C(213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066864
CAS Number: 80-59-1
EINECS: 201-295-0
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty