Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichloroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

A13922-18 A13922-30 A13922-30 A13922-18
AAA13922-18EA 46.29 USD InStock
CAAAA13922-18 AAA13922-18 CAAAA13922-30 AAA13922-30
3,5-Dichloroaniline 98%
3,5-Dichloroaniline


Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 259…260 °C/741 mm
Melting Pt: 49…53 °C
Density: 1.58
Flash Pt: 133 °C(271 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007774
CAS Number: 626-43-7
EINECS: 210-948-9
UN: 3442
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty