Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,10-Decanediol 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 1,10-Dihydroxy decane

A13923-22 A13923-22 A13923-36 A13923-36
AAA13923-36EA 185.99 USD InStock
AAA13923-36 CAAAA13923-22 CAAAA13923-36 AAA13923-22
1,10-Decanediol 97%
1,10-Decanediol


Formula: C₁₀H₂₂O₂
MW: 174.28 g/mol
Boiling Pt: 297 °C
Melting Pt: 71…74 °C
Density: 1.080
Flash Pt: 152 °C(306 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004749
CAS Number: 112-47-0
EINECS: 203-975-2
Merck Index: 13,02869
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty