Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-4-methoxybenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Amino-p-anisamide

A13942-14 A13942-06 A13942-14 A13942-06
AAA13942-06EA 55.29 USD InStock
CAAAA13942-14 AAA13942-06 AAA13942-14 CAAAA13942-06
3-Amino-4-methoxybenzamide 98%
3-Amino-4-methoxybenzamide


Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00017132
CAS Number: 17481-27-5
EINECS: 241-497-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty