Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropanemethylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (Aminomethyl)cyclopropane (Cyclopropylmethyl)amine

A13946-06 A13946-03 A13946-14 A13946-14 A13946-06 A13946-03
AAA13946-03EA 55.99 USD InStock
AAA13946-03 AAA13946-14 AAA13946-06 CAAAA13946-06 CAAAA13946-14 CAAAA13946-03
Cyclopropanemethylamine 98+%
Cyclopropanemethylamine


Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 85…86 °C
Density: 0.820
Flash Pt: 2 °C(36 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037147
CAS Number: 2516-47-4
EINECS: 219-737-6
UN: 2733
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty