Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Suberic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,6-Hexanedicarboxylic acid 1,8-Octanedioic acid

A13963-0E A13963-36 A13963-22 A13963-36 A13963-22 A13963-0E
AAA13963-22EA 56.79 USD InStock
CAAAA13963-22 CAAAA13963-36 CAAAA13963-0E AAA13963-22 AAA13963-0E AAA13963-36
Suberic acid 99%
Suberic acid


Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 230 °C/15 mm
Melting Pt: 141…144 °C
Density: 1,354 g/cm³ (21 °C)
Flash Pt: 203 °C(397 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004428
CAS Number: 505-48-6
EINECS: 208-010-9
Merck Index: 12,09031
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty