Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-4-nitrophenol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Hydroxy-5-nitroaniline

A13964-36 A13964-14 A13964-22 A13964-36 A13964-22 A13964-14
AAAA13964-36EA 622.67 USD InStock
CAAAA13964-22 AAAA13964-36 CAAAAA13964-36 CAAAA13964-14 AAA13964-14 AAA13964-22
2-Amino-4-nitrophenol 98%
2-Amino-4-nitrophenol


Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 141…145 °C
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007695
CAS Number: 99-57-0
EINECS: 202-767-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty