Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl pyruvate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Ketopropionic acid methyl ester 2-Oxopropionic acid methyl ester Pyruvic acid ethyl mester α-Ketopropionic acid methyl ester

A13966-22 A13966-14 A13966-22 A13966-36 A13966-36 A13966-14
AAA13966-22EA 106.5 USD InStock
AAA13966-22 CAAAA13966-22 CAAAA13966-14 CAAAAA13966-36 AAAA13966-36 AAA13966-14
Methyl pyruvate 98%
Methyl pyruvate


Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 135…137 °C
Density: 1.123
Flash Pt: 39 °C(102 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008754
CAS Number: 600-22-6
EINECS: 209-987-4
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty