Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Amino-2-methylquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-methylquinolin-6-amine

A13967-MD A13967-06 A13967-03 A13967-03 A13967-06
AAA13967-MDEA 53.89 USD InStock
CAAAA13967-03 CAAAA13967-06 AAA13967-MD AAA13967-06 AAA13967-03
6-Amino-2-methylquinoline 97%
6-Amino-2-methylquinoline


Formula: C₁₀H₁₀N₂
MW: 158.20 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00052600
CAS Number: 65079-19-8
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty