Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyrazine, (max. 1% H₂O) 99+% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

A13982-22 A13982-14 A13982-14 A13982-22
AAA13982-22EA 267.57 USD InStock
AAA13982-22 CAAAA13982-14 CAAAA13982-22 AAA13982-14
Pyrazine, (max. 1% H₂O) 99+% (dry weight)
Pyrazine


Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 113…116 °C
Melting Pt: 50…56 °C
Density: 1.031
Flash Pt: 55 °C(131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006122
CAS Number: 290-37-9
EINECS: 206-027-6
UN: 1325
ADR: "4.1",III
Merck Index: 13,08047
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty