Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentenoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Allylacetic acid

A13991-14 A13991-14 A13991-09 A13991-22 A13991-22 A13991-09
AAA13991-22EA 298.99 USD InStock
AAA13991-22 CAAAA13991-22 CAAAA13991-14 CAAAAA13991-09 AAA13991-14 AAAA13991-09
4-Pentenoic acid 98%
4-Pentenoic acid


Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 187…189 °C
Melting Pt: –22 °C
Density: 0.891
Flash Pt: 89 °C(192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004408
CAS Number: 591-80-0
EINECS: 209-732-7
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty