Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butylbenzoic acid 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-(tert-Butyl)benzoic acid

A13997-0B A13997-30 A13997-0B A13997-0I A13997-0I A13997-30
AAA13997-0BEA 104.99 USD InStock
CAAAA13997-0I AAA13997-0B CAAAA13997-0B AAA13997-30 AAA13997-0I CAAAA13997-30
4-tert-Butylbenzoic acid 99+%
4-tert-Butylbenzoic acid


Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 280 °C (dec.)
Melting Pt: 164…168 °C
Flash Pt: 180 °C(356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002563
CAS Number: 98-73-7
EINECS: 202-696-3
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty