Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,5-Pentanediol 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 1,5-Dihydroxy pentane

A13998-0E A13998-22 A13998-36 A13998-22 A13998-36 A13998-0E
AAA13998-22EA 52.24 USD InStock
AAA13998-22 CAAAA13998-36 CAAAA13998-0E AAA13998-36 AAA13998-0E CAAAA13998-22
1,5-Pentanediol 97%
1,5-Pentanediol


Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 241…242 °C
Melting Pt: –16 °C
Density: 0.992
Flash Pt: 129 °C(264 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002978
CAS Number: 111-29-5
EINECS: 203-854-4
Merck Index: 12,07256
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty