Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tropylium tetrafluoroborate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: TropyliumtetrafluoroborateCyclohepta-2,4,6-trienylium tetrafluoroborate

A14045-22 A14045-14 A14045-22 A14045-06 A14045-14 A14045-06
AAA14045-14EA 696.99 USD InStock
AAA14045-14 CAAAA14045-06 AAA14045-22 AAA14045-06 CAAAA14045-22 CAAAA14045-14
Tropylium tetrafluoroborate 97%
Tropylium tetrafluoroborate


Formula: C₇H₇BF₄
MW: 177.94 g/mol
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00013402
CAS Number: 27081-10-3
EINECS: 248-214-5
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty