Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chlorobenzylamine 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (3-Chlorophenyl)methanamine

A14078-09 A14078-09 A14078-18 A14078-18
AAA14078-09EA 83.15 USD InStock
AAA14078-09 AAA14078-18 CAAAA14078-18 CAAAA14078-09
3-Chlorobenzylamine 97+%
3-Chlorobenzylamine


Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 226 °C
Density: 1.159
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040752
CAS Number: 4152-90-3
EINECS: 223-985-0
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty