Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminothiophenol 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Aminobenzenethiol 4-Aminophenyl mercaptan 4-Mercaptoaniline

A14082-14 A14082-22 A14082-06 A14082-14 A14082-22 A14082-06
AAA14082-14EA 198.99 USD InStock
AAA14082-14 CAAAA14082-06 CAAAA14082-14 CAAAA14082-22 AAA14082-06 AAA14082-22
4-Aminothiophenol 97%
4-Aminothiophenol


Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 156…157 °C
Melting Pt: 37…42 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007880
CAS Number: 1193-02-8
EINECS: 214-763-4
UN: 3263
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty