Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butylthiazol-2-ylamine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-4-(tert-butyl)-1,3-thiazole

A14115-06 A14115-03 A14115-06 A14115-03 A14115-14 A14115-14
AAA14115-03EA 33.49 USD InStock
CAAAA14115-03 AAA14115-03 CAAAA14115-14 AAA14115-14 CAAAA14115-06 AAA14115-06
4-tert-Butylthiazol-2-ylamine 98%
4-tert-Butylthiazol-2-ylamine


Formula: C₇H₁₂N₂S
MW: 156.25 g/mol
Melting Pt: 98…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00051644
CAS Number: 74370-93-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty