Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopentanecarbonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cyclopentanecarboxylic acid chloride Cyclopentanecarboxylic chloride

A14142-14 A14142-06 A14142-22 A14142-06 A14142-22 A14142-14
AAA14142-14EA 228.99 USD InStock
AAA14142-14 AAA14142-22 CAAAA14142-06 CAAAA14142-22 CAAAA14142-14 AAA14142-06
Cyclopentanecarbonyl chloride 98%
Cyclopentanecarbonyl chloride


Formula: C₆H₉ClO
MW: 132.59 g/mol
Boiling Pt: 160…162 °C
Density: 1.091
Flash Pt: 60 °C(140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001370
CAS Number: 4524-93-0
EINECS: 224-856-1
UN: 2920
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty