Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Aza-2-thiothymine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 6-Methyl-3-thioxo-3,4-dihydro-1,2,4-triazin-5(2H)-one

A14167-06 A14167-22 A14167-14 A14167-14 A14167-06 A14167-22
AAA14167-06EA 120.9 USD InStock
AAA14167-06 AAA14167-14 AAA14167-22 CAAAA14167-22 CAAAA14167-06 CAAAA14167-14
6-Aza-2-thiothymine 98%
6-Aza-2-thiothymine


Formula: C₄H₅N₃OS
MW: 143.17 g/mol
Melting Pt: 218…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00006458
CAS Number: 615-76-9
EINECS: 210-445-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty