Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hexylbenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A14168-06 A14168-03 A14168-06 A14168-03
AAA14168-06EA 205.99 USD InStock
AAA14168-06 AAA14168-03 CAAAA14168-03 CAAAA14168-06
4-Hexylbenzoic acid 99%
4-Hexylbenzoic acid


Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 97…99 °C (C-Nematic)
Storage Temperature: Ambient
MDL Number: MFCD00013997
CAS Number: 21643-38-9
EINECS: 244-490-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty