Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-3,5-dimethylisoxazole 97%

Supplier: Thermo Scientific Chemicals
Warning

A14179-03 A14179-14 A14179-06 A14179-06 A14179-14 A14179-03
AAA14179-06EA 201.99 USD InStock
AAA14179-06 AAA14179-14 AAA14179-03 CAAAA14179-03 CAAAA14179-14 CAAAA14179-06
4-Bromo-3,5-dimethylisoxazole 97%
4-Bromo-3,5-dimethylisoxazole


Formula: C₅H₆BrNO
Boiling Pt: 176 °C
Density: 1,49
Flash Pt: 73 °C(163 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00068187
CAS Number: 10558-25-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty