Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Phenylglycine 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-(Phenylamino)acetic acid2-Anilinoacetic acid

A14182-18 A14182-18 A14182-09 A14182-09
AAA14182-09EA 36.51 USD InStock
AAA14182-09 AAA14182-18 CAAAA14182-09 CAAAA14182-18
N-Phenylglycine 97%
N-Phenylglycine
Applications: Pharmaceutical intermediate.


Formula: C₈H₉NO₂
MW: 151.17 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00014009
CAS Number: 103-01-5
EINECS: 203-070-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty