Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Phenyl-2-butenal mixture E/Z 97%

Supplier: Thermo Scientific Chemicals
Warning

A14185-14 A14185-06 A14185-14 A14185-06
AAA14185-14EA 176.99 USD InStock
AAA14185-14 AAA14185-06 CAAAA14185-14 CAAAA14185-06
2-Phenyl-2-butenal mixture E/Z 97%
2-Phenyl-2-butenal


Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 115 °C/15 mm
Storage Temperature: Ambient
MDL Number: MFCD00053158
CAS Number: 4411-89-6
EINECS: 224-567-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty