Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Glutamic acid 99+%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid (R)-(-)-2-Aminopentanedioic acid (R)-(-)-Glutamic acid (R)-(-)-α-Glutamic acid D(-)-1-Aminopropane-1,3-dicarboxylic acid D(-)-2-Aminopentanedioic acid D(-)-α-Glutamic acid H-D-Glu-OH

A14191-14 A14191-22 A14191-06 A14191-22 A14191-14 A14191-06
AAAA14191-22EA 464.99 USD InStock
AAAA14191-22 CAAAA14191-06 CAAAAA14191-22 AAA14191-06 AAA14191-14 CAAAA14191-14
D-(-)-Glutamic acid 99+%
D(-)-Glutamic acid


Formula: C₅H₉NO₄
MW: 147.13 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 199 °C (dec.)
Density: 1.533 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063112
CAS Number: 6893-26-1
EINECS: 230-000-8
Merck Index: 13,04482
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty