Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylenebis(triphenylphosphonium bromide) 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14214-18 A14214-09 A14214-09 A14214-18
AAA14214-18EA 184.37 USD InStock
CAAAA14214-09 AAA14214-18 CAAAA14214-18 AAA14214-09
Tetramethylenebis(triphenylphosphonium bromide) 98+%
Tetramethylenebis(triphenylphosphonium bromide)


Formula: C₄₀H₃₈Br₂P₂
Melting Pt: 292…293 °C
Storage Temperature: Ambient
MDL Number: MFCD00031714
CAS Number: 15546-42-6
EINECS: 239-598-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty